Anti-BSG Mouse Monoclonal Antibody (Purified) [clone: CB43]
Proveedor: Biotium
BNUB0363-500 BNUB0363-100
BNUB0363-500EA 511 EUR
BNUB0363-500 BNUB0363-100
Anti-BSG Mouse Monoclonal Antibody (Purified) [clone: CB43]
Anticuerpos
CD147 is a transmembrane glycoprotein of the immunoglobulin superfamily. It is expressed more intensely on thymocytes than on mature peripheral blood T cells. CD147 is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is involved in the regulation of matrix remodeling at the epidermal-dermal interface. It stimulates the production of interstitial collagenase, gelatinase A, stromelysin-1 and various metalloproteinases (MMPs) by fibroblasts. These enzymes, which are typically increased during tissue degradation and wound healing, are important factors in cancer invasion and metastasis.
Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: CB43
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human