Anti-TNC Mouse Monoclonal Antibody (CF647) [clone: T2H5]

Proveedor: Biotium
BNC470392-500 BNC470392-100
BNC470392-500EA 560 EUR
BNC470392-500 BNC470392-100
Anti-TNC Mouse Monoclonal Antibody (CF647) [clone: T2H5]
Anticuerpos
In Western blotting, this antibody reacts with two bands of ~MW of 210 kDa and 300 kDa, identified as two isoforms of Tenascin C. Specificity of this MAb is validated by sequential immunoprecipitation with a PAb against Tenascin C. Tenascin C is a multifunctional, disulfide-linkedhexameric extracellular matrix glycoprotein expressed in association with mesenchymal epithelial interactions during development and in the neo-vasculature and stroma of undifferentiated tumors. In adults, it is restricted to certain epithelial-stromal interfaces and increases markedly in hyper-proliferativeMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno TNC
Environmentally Preferable
Nombre del antígeno Tenascin C
Sinónimos de antígenos Neuronectin|GP 150-225|Cytotactin|Myotendinous antigen|Hexabrachion (HXB)|JI|GMEM|TNC|Glioma-associated-extracellular matrix antigen
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 647
Clon Clone T2H5
Reactividad Human
Huésped Mouse
ID del gen 143250
Isotipo IgG1 kappa
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR