Anti-ESR1 Mouse Monoclonal Antibody (CF594) [clone: AER314]
Proveedor: Biotium
BNC940406-500 BNC940406-100
BNC940406-500EA 560 EUR
BNC940406-500 BNC940406-100
Anti-ESR1 Mouse Monoclonal Antibody (CF594) [clone: AER314]
Anticuerpos
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. Epitope of this MAb is mapped between aa120-170. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.
Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: AER314
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow, Human, Mouse, Rabbit, Rat