Anti-TNFRSF8 Mouse Monoclonal Antibody (CF594) [clone: CD30/412]

Proveedor: Biotium
BNC940412-500 BNC940412-100
BNC940412-500EA 560 EUR
BNC940412-500 BNC940412-100
Anti-TNFRSF8 Mouse Monoclonal Antibody (CF594) [clone: CD30/412]
Anticuerpos
Recognizes a single chain glycoprotein of 105/120 kDa, identified as CD30/Ki-1. CD30 is synthesized as a 90 kDa precursor, which is processed in the Golgi complex into a membrane-bound phosphorylated mature 105/120 kDa glycoprotein. In Hodgkin's disease, CD30/Ki-1 antigen is expressed by mononuclear-Hodgkin and multinucleated Reed-Sternberg cells. It is also expressed by the tumor cells of a majority of anaplastic large cell lymphomas as well as by a varying proportion of activated T and B cells. This MAb distinguishes large cell lymphomas derived from activated lymphoid cells from histiocytic malignancies and lymphomas derived from resting and precursor lymphoid cells or from anaplastic carcinomas. About one third of the Ki-1 positive lymphomas lack the leukocyte common antigen (CD45).

Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: CD30/412
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR