Anti-ESR1 Mouse Monoclonal Antibody (CF568) [clone: ESR1/420]

Proveedor: Biotium
BNC680420-500 BNC680420-100
BTIUBNC680420-500EA 560 EUR
BTIUBNC680420-500 BTIUBNC680420-100
Anti-ESR1 Mouse Monoclonal Antibody (CF568) [clone: ESR1/420]
Anticuerpos
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. Epitope of this MAb is mapped between aa300-550. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ESR1/420
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR