Anti-BSG Mouse Monoclonal Antibody (CF647) [clone: 8D6]

Proveedor: Biotium
BNC470424-500 BNC470424-100
BNC470424-500EA 560 EUR
BNC470424-500 BNC470424-100
Anti-BSG Mouse Monoclonal Antibody (CF647) [clone: 8D6]
Anticuerpos
This MAb recognizes extracellular epitope 2 within the N-terminal Ig domain of human CD147. It is expressed more intensely on thymocytes than on mature peripheral blood T cells. CD147 is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. It stimulates the production of interstitial collagenase, gelatinase A, stromelysin-1 and various metalloproteinases (MMPs) by fibroblasts. These enzymes are important factors in cancer invasion and metastasis.

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: 8D6
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR