Anti-S100A9 Mouse Monoclonal Antibody (Purified) [clone: CAGB/426]

Proveedor: Biotium
BNUB0426-500 BNUB0426-100
BNUB0426-500EA 511 EUR
BNUB0426-500 BNUB0426-100
Anti-S100A9 Mouse Monoclonal Antibody (Purified) [clone: CAGB/426]
Anticuerpos
Recognizes a 12-14 kDa doublet of Calgranulin A/B (also known as S100A8/A9 or MRP-8/14); expressed by granulocytes, monocytes and by tissue macrophages.The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. Altered expression of this protein is associated with the disease cystic fibrosis. This MAb reacts with neutrophils, monocytes, macrophages, and squamous mucosal epithelia and has been shown as an important marker for identifying macrophages in tissue sections.

Type: Primary
Antigen: S100A9
Clonality: Monoclonal
Clone: CAGB/426
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR