Anti-B2M Mouse Monoclonal Antibody (CF568) [clone: C21.48A1]

Proveedor: Biotium
BNC680433-500 BNC680433-100
BNC680433-500EA 560 EUR
BNC680433-500 BNC680433-100
Anti-B2M Mouse Monoclonal Antibody (CF568) [clone: C21.48A1]
Anticuerpos
The antibody recognizes the hidden determinant of β-2 microglobulin (i.e. binding to its determinant is available only when the chain is separated from the HLA heavy chain. β-2 microglobulin is a 12 kDa protein with a pI of 5.6. Serum β2 microglobulin levels are a reflection of cell turnover. Levels rise with fever, inflammation, and infection. Increased serum levels are also seen in B-cell malignancies and in renal failure and may indicate a worse prognosis for patients with early-stage Hodgkin's lymphoma. In urine, increased levels are seen in proximal renal tubular disease as well asMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno B2M B2M
Environmentally Preferable
Nombre del antígeno Beta-2-Microglobulin Beta-2-Microglobulin
Sinónimos de antígenos Hdcma22p|Beta 2 microglobulin|Beta chain of MHC class I molecules|B2M|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin Hdcma22p|Beta 2 microglobulin|Beta chain of MHC class I molecules|B2M|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin
Tipo de anticuerpo Primario Primario
Clonalidad Monoclonal Monoclonal
Conjugado CF® 568 CF® 568
Clon Clone C21.48A1 Clone C21.48A1
Reactividad Primates,Human Primates,Human
Huésped Mouse Mouse
ID del gen 534255 534255
Isotipo IgG2b kappa IgG2b kappa
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR