Anti-SILV Mouse Monoclonal Antibody (Biotin) [clone: HMB45]

Proveedor: Biotium
BNCB0444-500 BNCB0444-100
BNCB0444-500EA 560 EUR
BNCB0444-500 BNCB0444-100
Anti-SILV Mouse Monoclonal Antibody (Biotin) [clone: HMB45]
Anticuerpos
By immunohistochemistry, this antibodyspecifically recognizes a protein in melanocytes and melanomas. This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin. Metastatic amelanotic melanoma can often be confused with a variety of poorly differentiated carcinomas, large cell lymphomas, and sarcomas using H & E stains alone. It is also difficult to differentiate melanoma fromMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno SILV
Nombre del antígeno Pmel17 / gp100 / SILV
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Biotin
Clon Clone HMB45
Reactividad Human
Huésped Mouse
ID del gen 95972
Isotipo IgG1 kappa
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR