Anti-SILV Mouse Monoclonal Antibody (CF405S) [clone: HMB45]

Proveedor: Biotium
BNC040444-500 BNC040444-100
BNC040444-500EA 560 EUR
BNC040444-500 BNC040444-100
Anti-SILV Mouse Monoclonal Antibody (CF405S) [clone: HMB45]
Anticuerpos
By immunohistochemistry, this antibodyspecifically recognizes a protein in melanocytes and melanomas. This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin. Metastatic amelanotic melanoma can often be confused with a variety of poorly differentiated carcinomas, large cell lymphomas, and sarcomas using H & E stains alone. It is also difficult to differentiate melanoma fromMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno SILV
Nombre del antígeno Pmel17 / gp100 / SILV
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 405S
Clon Clone HMB45
Reactividad Human
Huésped Mouse
ID del gen 95972
Isotipo IgG1 kappa
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR