Anti-ESR2 Mouse Monoclonal Antibody (Purified) [clone: ERb455]
Proveedor: Biotium
BNUB0455-500 BNUB0455-100
BNUB0455-500EA 511 EUR
BNUB0455-500 BNUB0455-100
Anti-ESR2 Mouse Monoclonal Antibody (Purified) [clone: ERb455]
Anticuerpos
Estrogen receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of ligand-activated transcription factors. Estrogen receptors, including ER-alpha and ER-beta, contain DNA binding and ligand binding domains and are critically involved in regulating the normal function of reproductive tissues. They are located in the nucleus, though some estrogen receptors associate with the cell surface membrane and can be rapidly activated by exposure of cells to estrogen. ER-alpha and ER-beta are differentially activated by various ligands. Receptor-ligand interactions trigger a cascade of events, including dissociation from heat shock proteins, receptor dimerization, phosphorylation and the association of the hormone activated receptor with specific regulatory elements in target genes. Evidence suggests that ER-alpha and ER-beta may be regulated by distinct mechanisms even though they share many functional characteristics.
Type: Primary
Antigen: ESR2
Clonality: Monoclonal
Clone: ERb455
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Horse, Human, Monkey, Mouse, Pig, Rat, Sheep