Anti-ESR1 Mouse Monoclonal Antibody (Biotin) [clone: ER505]

Proveedor: Biotium
BNCB0505-500 BNCB0505-100
BTIUBNCB0505-500EA 560 EUR
BTIUBNCB0505-500 BTIUBNCB0505-100
Anti-ESR1 Mouse Monoclonal Antibody (Biotin) [clone: ER505]
Anticuerpos
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. This MAb is excellent for staining of formalin-fixed, paraffin-embedded breast carcinomas.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ER505
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR