Anti-ESR1 Mouse Monoclonal Antibody (Purified) [clone: ER506]

Proveedor: Biotium
BNUB0506-500 BNUB0506-100
BTIUBNUB0506-500EA 511 EUR
BTIUBNUB0506-500 BTIUBNUB0506-100
Anti-ESR1 Mouse Monoclonal Antibody (Purified) [clone: ER506]
Anticuerpos
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. It strongly stains nuclei of epithelial cells in breast carcinomas.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ER506
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno ESR1
Nombre del antígeno Estrogen Receptor
Environmentally Preferable
Sinónimos de antígenos Nuclear receptor subfamily 3 group A member 1|Estrogen Receptor alpha delta 4*5,6,7*/654 isoform|Estrogen Receptor alpha delta 4 49 isoform
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon Clone ER506
Reactividad Human
Huésped Mouse
ID del gen 208124
Isotipo IgG1 kappa
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,2 mg/ml
Storage buffer PBS/0,05% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR