Anti-ESR1 Mouse Monoclonal Antibody (CF405S) [clone: ER506]

Proveedor: Biotium
BNC040506-500 BNC040506-100
BTIUBNC040506-500EA 560 EUR
BTIUBNC040506-500 BTIUBNC040506-100
Anti-ESR1 Mouse Monoclonal Antibody (CF405S) [clone: ER506]
Anticuerpos
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. It strongly stains nuclei of epithelial cells in breast carcinomas.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ER506
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR