Anti-CD7 Mouse Monoclonal Antibody (Purified) [clone: C7/511]
Proveedor: Biotium
BNUB0511-500 BNUB0511-100
BNUB0511-500EA 511 EUR
BNUB0511-500 BNUB0511-100
Anti-CD7 Mouse Monoclonal Antibody (Purified) [clone: C7/511]
Anticuerpos
Recognizes a protein of 40 kDa, identified as CD7, a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxyl-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3'-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation.
Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: C7/511
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human