Anti-CD68 Mouse Monoclonal Antibody (CF568) [clone: KP1]

Proveedor: Biotium
BNC680512-500 BNC680512-100
BNC680512-500EA 560 EUR
BNC680512-500 BNC680512-100
Anti-CD68 Mouse Monoclonal Antibody (CF568) [clone: KP1]
Anticuerpos
This antibody recognizes a glycoprotein of 110 kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia andMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno CD68 CD68
Environmentally Preferable
Sinónimos de antígenos Scavenger Receptor Class D Member-1|Microsialin|GP110|SCARD1|LAMP4|Macrosialin Scavenger Receptor Class D Member-1|Microsialin|GP110|SCARD1|LAMP4|Macrosialin
Tipo de anticuerpo Primario Primario
Clonalidad Monoclonal Monoclonal
Conjugado CF® 568 CF® 568
Clon Clone KP1 Clone KP1
Reactividad Rabbit,Monkey,Human,Cat Rabbit,Monkey,Human,Cat
Huésped Mouse Mouse
ID del gen 647419 647419
Isotipo IgG1 kappa IgG1 kappa
InmunoQuímica Yes Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR