Anti-CD14 Mouse Monoclonal Antibody (CF640R) [clone: LPSR/553]

Proveedor: Biotium
BNC400553-500 BNC400553-100
BNC400553-500EA 560 EUR
BNC400553-500 BNC400553-100
Anti-CD14 Mouse Monoclonal Antibody (CF640R) [clone: LPSR/553]
Anticuerpos
Recognizes a protein of 55 kDa, identified as CD14 (also known lipopolysaccharide receptor). CD14 is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: LPSR/553
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR