Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: VU-2G7]

Proveedor: Biotium
BNC940630-500 BNC940630-100
BNC940630-500EA 560 EUR
BNC940630-500 BNC940630-100
Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: VU-2G7]
Anticuerpos
MAb VU-2G7 reacts with MUC1, a large transmembrane glycoprotein expressed on the ductal surface of normal glandular epithelia. The dominant epitope of VU-2G7 has not been established with epitope fingerprinting. The extracellular domain of MUC1 largely consists of a highly conserved, O-glycosylated 20 amino acids tandem repeat which can occur 30-100 times per molecule depending on the length of the allele involved. In the vast majority of human carcinomas this protein is up regulated and poorly glycosylated and appears on the cell surface in a non-polarized fashion.

Type: Primary
Antigen:More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno MUC1
Environmentally Preferable
Nombre del antígeno Mucin 1 / EMA / Episialin / CD227
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 594
Clon Clone VU-2G7
Reactividad Human
Huésped Mouse
ID del gen 89603
Isotipo IgG1 kappa
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR