Anti-IGKC Mouse Monoclonal Antibody (CF488A) [clone: TB28-2]

Proveedor: Biotium
BNC880708-500 BNC880708-100
BNC880708-500EA 560 EUR
BNC880708-500 BNC880708-100
Anti-IGKC Mouse Monoclonal Antibody (CF488A) [clone: TB28-2]
Anticuerpos
This MAb is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. It recognizes human Ig kappa light chains of both secreted and cell surface immunoglobulin. It detects also free kappa light chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the kappa light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant.

Type: Primary
Antigen: IGKC
Clonality: Monoclonal
Clone: TB28-2
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR