Anti-HSPD1 Mouse Monoclonal Antibody (CF405S) [clone: GROEL/730]

Proveedor: Biotium
BNC040730-500 BNC040730-100
BTIUBNC040730-500EA 560 EUR
BTIUBNC040730-500 BTIUBNC040730-100
Anti-HSPD1 Mouse Monoclonal Antibody (CF405S) [clone: GROEL/730]
Anticuerpos
Recognizes a 60 kDa protein, identified as the heat shock protein 60 (hsp60). A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp), more appropriately called as stress response proteins (srp). Hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial hsp60, but also against its mammalian homolog. CloneMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno HSPD1
Environmentally Preferable
Nombre del antígeno HSP60
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 405S
Clon Clone GROEL/730
Reactividad Human
Huésped Mouse
ID del gen 595053
Isotipo IgG1 kappa
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR