Anti-MUC1 Mouse Monoclonal Antibody (CF647) [clone: GP1.4 E29]

Proveedor: Biotium
BNC470743-500 BNC470743-100
BNC470743-500EA 560 EUR
BNC470743-500 BNC470743-100
Anti-MUC1 Mouse Monoclonal Antibody (CF647) [clone: GP1.4 E29]
Anticuerpos
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. The α subunit has cell adhesive properties. It can act both as an adhesion and an anti-adhesion protein. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. The β subunit contains a C-terminal domain, which is involved in cell signaling, through phosphorylations and protein-protein interactions. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinoma tissues. Antibody to EMA is useful as a pan-More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno MUC1 MUC1
Environmentally Preferable
Nombre del antígeno Mucin 1 / EMA / Episialin / CD227 Mucin 1 / EMA / Episialin / CD227
Tipo de anticuerpo Primario Primario
Clonalidad Monoclonal Monoclonal
Conjugado CF® 647 CF® 647
Clon Clone GP1.4 E29 Clone GP1.4 E29
Reactividad Human Human
Huésped Mouse Mouse
ID del gen 89603 89603
Isotipo IgGs IgGs
InmunoQuímica Yes Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR