Anti-CD66 molecule Mouse Monoclonal Antibody [clone: COL-1 CEA31 C66/261]

Proveedor: Biotium
BNUB0747-500 BNUB0747-100
BNUB0747-500EA 511 EUR
BNUB0747-500 BNUB0747-100
Anti-CD66 molecule Mouse Monoclonal Antibody [clone: COL-1 CEA31 C66/261]
Anticuerpos
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci of gastric carcinoma and in distinguishing pulmonary adenocarcinomas (60-70% are CEA ) from pleural mesotheliomas (rarely or weakly CEA ). Anti-CEA positivity is seen in adenocarcinomas from the lung, colon, stomach, esophagus, pancreas, gallbadder, urachus, salivary gland, ovary, and endocervix.

  • Purified

Type: Primary
Antigen: CD66 molecule
Clonality: Monoclonal
Clone: COL-1 CEA31 C66/261
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR