Anti-CD8A Mouse Monoclonal Antibody (CF647) [clone: C8/144B]

Proveedor: Biotium
BNC470749-500 BNC470749-100
BNC470749-500EA 560 EUR
BNC470749-500 BNC470749-100
Anti-CD8A Mouse Monoclonal Antibody (CF647) [clone: C8/144B]
Anticuerpos
CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 β chain (CD8 α/β) or as a homodimer (CD8 α/α). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. For mature T-cells, CD4 and CD8 are mutually exclusive, so anti-CD8, generally used in conjunction with anti-CD4. It is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral T-cell lymphomas areMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno CD8A
Nombre del antígeno CD8
Environmentally Preferable
Sinónimos de antígenos T8/Leu-2 T-lymphocyte differentiation antigen|LYT3|alpha polypeptide (p32)|T-cell surface glycoprotein CD8 alpha chain|CD8 antigen|MAL|Ly3
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 647
Clon Clone C8/144B
Reactividad Human
Huésped Mouse
ID del gen 85258
Isotipo IgG1 kappa
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR