Anti-BCL2L1 Mouse Monoclonal Antibody [clone: BX006 2H12]
Proveedor: Biotium
BNUB0752-500 BNUB0752-100
BNUB0752-500EA 511 EUR
BNUB0752-500 BNUB0752-100
Anti-BCL2L1 Mouse Monoclonal Antibody [clone: BX006 2H12]
Anticuerpos
Recognizes a protein of 27 kDa, identified as the Bcl-X protein. This MAb shows no cross-reaction with Bcl-2 or Bax protein. Bcl-X has two isoforms, Bcl-XL (long), a 241 amino acid protein which suppresses cell death. And Bcl-XS (short), a 178 amino acid protein lacking a 63 amino acid domain which functions as a dominant inhibitor of Bcl-2. This MAb reacts with both Bcl-XS and Bcl-XL proteins.
  • Purified
Type: Primary
Antigen: BCL2L1
Clonality: Monoclonal
Clone: BX006 2H12
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Mouse, Pig, Rat