Anti-CA9 Mouse Monoclonal Antibody (CF594) [clone: CA9/781]

Proveedor: Biotium
BNC940781-500 BNC940781-100
BNC940781-500EA 560 EUR
BNC940781-500 BNC940781-100
Anti-CA9 Mouse Monoclonal Antibody (CF594) [clone: CA9/781]
Anticuerpos
Recognizes a glycoprotein of ~200 kDa, identified as carbonic anhydrase IX (CAIX/gp200). Carbonic Anhydrases (CAs) are members of a large family of zinc metallo-enzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption and the formation of aqueous humor, cerebrospinal fluid, saliva and gastric juice. They show extensive diversity in distribution and in their subcellular localization. CA IX is specifically expressed in clear-cell renal carcinomas.

Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: CA9/781
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Horse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR