Anti-Phosphotyrosine Mouse Monoclonal Antibody (CF405S) [clone: PY793]

Proveedor: Biotium
BNC040793-500 BNC040793-100
BNC040793-500EA 560 EUR
BNC040793-500 BNC040793-100
Anti-Phosphotyrosine Mouse Monoclonal Antibody (CF405S) [clone: PY793]
Anticuerpos
Protein phosphorylation is a fundamental event in the regulation of a large number of intracellular processes. Phosphorylation of specific tyrosine residues is the result of activation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be auto-phosphorylated kinases or certain cellular protein substrates. Tyrosine-phosphorylated proteins are involved in signal transduction and in the regulation of cell proliferation. Antibody to phosphotyrosine provides an excellent tool for the detection, characterization, and purification of phosphotyrosineMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno pY
Environmentally Preferable
Nombre del antígeno Phosphotyrosine
Sinónimos de antígenos P-Tyr
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 405S
Clon Clone PY793
Reactividad Multiple
Huésped Mouse
Isotipo lgG2b
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR