Anti-NCAM1 Mouse Monoclonal Antibody (CF568) [clone: NCAM1/795]

Proveedor: Biotium
BNC680795-500 BNC680795-100
BNC680795-500EA 560 EUR
BNC680795-500 BNC680795-100
Anti-NCAM1 Mouse Monoclonal Antibody (CF568) [clone: NCAM1/795]
Anticuerpos
This MAb reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Three isoforms of neural cell adhesion molecule (NCAM) are produced by differential splicing of the RNA transcript from a single gene. The 135 kDa isoform is the basic molecule, which is glycosylated or sialylated to produce the mature species. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas). It is alsoMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno NCAM1
Nombre del antígeno Neural Cell Adhesion Molecule 1
Environmentally Preferable
Sinónimos de antígenos NCAM120|NCAM|NCAM180|Leu-19|MSK39|NKH1|NCAM140|Neural Cell Adhesion Molecule
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 568
Clon Clone NCAM1/795
Reactividad Human
Huésped Mouse
ID del gen 503878
Isotipo IgG1 kappa
Western blot Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR