Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: KRT19/799]

Proveedor: Biotium
BNCB0799-500 BNCB0799-100
BNCB0799-500EA 560 EUR
BNCB0799-500 BNCB0799-100
Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: KRT19/799]
Anticuerpos
Recognizes a protein of 40 kDa, identified as cytokeratin-19 (CK19), which is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.

Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: KRT19/799
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR