Anti-KLK3 Mouse Monoclonal Antibody (CF488A) [clone: KLK3/801]

Proveedor: Biotium
BNC880801-500 BNC880801-100
BNC880801-500EA 560 EUR
BNC880801-500 BNC880801-100
Anti-KLK3 Mouse Monoclonal Antibody (CF488A) [clone: KLK3/801]
Anticuerpos
Recognizes a single protein of 33-34 kDa, identified as the prostate specific antigen (PSA). This MAb is highly specific to PSA and stains prostatic secretory and ductal epithelium in both normal and neoplastic tissues. PSA is a chymotrypsin-like serine protease (kallikrein family) exclusively produced by the prostate epithelium, and abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT inMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno KLK3
Environmentally Preferable
Nombre del antígeno PSA
Sinónimos de antígenos Seminin|P-30 antigen|Semenogelase|prostate-specific (APS)|Kallikrein-3 (KLK3)|Gamma-seminoprotein|antigen|hK3|Kallikrein related peptidase 3|KLK2A1
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 488A
Clon Clone KLK3/801
Reactividad Human
Huésped Mouse
ID del gen 171995
Isotipo IgG1 kappa
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR