Anti-TP53 Mouse Monoclonal Antibody (CF647) [clone: TRP/817]

Proveedor: Biotium
BNC470817-500 BNC470817-100
BNC470817-500EA 560 EUR
BNC470817-500 BNC470817-100
Anti-TP53 Mouse Monoclonal Antibody (CF647) [clone: TRP/817]
Anticuerpos
Recognizes a 53 kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53 protein. p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to MDM2, SV40 T antigen and human papilloma virus E6 protein. Positive nuclear staining with p53 antibody has been reported to be a negative prognostic factor in breast carcinoma, lung carcinoma, colorectal, and urothelial carcinoma. Anti-p53 positivity has also been used to differentiate uterine serous carcinoma from endometrioid carcinoma as well as to detect intratubular germ cell neoplasia. Mutations involving p53 are found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: TRP/817
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR