Anti-CD79A Mouse Monoclonal Antibody (CF594) [clone: JCB117 HM47/A9]

Proveedor: Biotium
BNC940828-500 BNC940828-100
BNC940828-500EA 560 EUR
BNC940828-500 BNC940828-100
Anti-CD79A Mouse Monoclonal Antibody (CF594) [clone: JCB117 HM47/A9]
Anticuerpos
A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells. This complex of mb-1 and B29 polypeptides and immunoglobulin constitute the B cell Ag receptor. CD79a first appears at pre B cell stage, early in maturation, and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno CD79A
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 594
Clon Clone JCB117 HM47/A9
Reactividad Pig,Monkey,Human,Bovine,Rat,Mouse
Huésped Mouse
ID del gen 631567
Isotipo IgGs
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR