Anti-KRT10 Mouse Monoclonal Antibody (Purified) [clone: KRT10/844]

Proveedor: Biotium
BNUB0844-500 BNUB0844-100
BNUB0844-500EA 511 EUR
BNUB0844-500 BNUB0844-100
Anti-KRT10 Mouse Monoclonal Antibody (Purified) [clone: KRT10/844]
Anticuerpos
This MAb recognizes a protein of 56.5 kDa, identified as cytokeratin 10 (CK10). CK10 is expressed in all suprabasal layers of the epidermis. In the epidermis, expression of CK10 strictly parallels the extent of differentiation; it is absent in the basal layer, appears in the first suprabasal layers and increases in concentration towards the granular layer. However, CK10 is rarely detected in early stages of vulvar squamous carcinomas (tumors less than 2 cm, clinical stage I) regardless of the tumor grade. In larger and more advanced tumors (greater than 2 cm, clinical stages II and III), CK10 is detected very frequently. Expression of CK10 is related to maturation of malignant keratinocytes, being preferentially detected in more-differentiated parts.

Type: Primary
Antigen: KRT10
Clonality: Monoclonal
Clone: KRT10/844
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR