Anti-RBP1 Mouse Monoclonal Antibody (Biotin) [clone: G4E4]
Proveedor: Biotium
BNCB0855-500 BNCB0855-100
BNCB0855-500EA 560 EUR
BNCB0855-500 BNCB0855-100
Anti-RBP1 Mouse Monoclonal Antibody (Biotin) [clone: G4E4]
Anticuerpos
This MAb recognizes an epitope within the 74-182 C-terminal sequence (11kD peptide fragment) of human serum Cellular Retinol Binding Protein 1 (CRBP 1), a single-chain glycoprotein belonging to the superfamily of hydrophobic molecule transporter proteins, which is responsible for transport of retinol (vitamin A1) from the liver to peripheral target tissues, like the eye, where it mediates the cellular uptake. CRBP 1 is synthesized by hepatic parenchymal cells where it becomes bound to its ligand retinol and is then released into the circulation, where it binds further to the proteinMore Product Information