Anti-ALPP Mouse Monoclonal Antibody (CF647) [clone: ALPP/870]

Proveedor: Biotium
BNC470870-500 BNC470870-100
BNC470870-500EA 560 EUR
BNC470870-500 BNC470870-100
Anti-ALPP Mouse Monoclonal Antibody (CF647) [clone: ALPP/870]
Anticuerpos
Reacts with a 70 kDa membrane-bound isozyme (Regan and Nagao type) of Placental Alkaline Phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. It is highly specific for PLAP and shows no cross-reaction with other isozymes of alkaline phosphatase. Anti-PLAP reacts with germ cell tumors and can discriminate between these and other neoplasms. Somatic neoplasms e.g. breast, gastrointestinal, prostatic, and urinary cancers may also immunoreact with antibodies to PLAP. Anti-PLAP positivity in conjunction with anti-keratin negativity favors seminoma over carcinoma. Germ cell tumors are usually anti-keratin positive, but they regularly fail to stain with anti-EMA, whereas most carcinomas stain with anti-EMA. Anti-PLAP has been useful in the diagnosis of gestational trophoblastic disease.

Type: Primary
Antigen: ALPP
Clonality: Monoclonal
Clone: ALPP/870
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR