Anti-MUC5AC Mouse Monoclonal Antibody (Biotin) [clone: CLH2]

Proveedor: Biotium
BNCB0897-500 BNCB0897-100
BNCB0897-500EA 560 EUR
BNCB0897-500 BNCB0897-100
Anti-MUC5AC Mouse Monoclonal Antibody (Biotin) [clone: CLH2]
Anticuerpos
Mucin 5AC glycoprotein (MUC5AC) is a HMW glycoprotein belonging to the superfamily of mucins. Mucins are produced by epithelial cells and can be divided into two families; secretory mucins and membrane bound mucins. MUC5AC is a mucus-forming, secreted mucin that is found in normal gastric and tracheo-bronchial mucosa, but absent from normal colon. MUC5AC expression is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-MUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastatic to the ovary. MUC5AC antibodies may also be useful for identification of intestinal metaplasia as well as in the identification of pancreatic carcinoma and pre-cancerous changes vs. normal pancreas.

Type: Primary
Antigen: MUC5AC
Clonality: Monoclonal
Clone: CLH2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR