Anti-FCGR3A Mouse Monoclonal Antibody (Purified) [clone: HuNK2]

Proveedor: Biotium
BNUB0928-500 BNUB0928-100
BNUB0928-500EA 511 EUR
BNUB0928-500 BNUB0928-100
Anti-FCGR3A Mouse Monoclonal Antibody (Purified) [clone: HuNK2]
Anticuerpos
Recognizes a protein of 50-65 kDa, identified as CD16 (Workshop IV; Code N39 ) (also known low affinity Fc receptor III for IgG (FcRIII) or Leu 11). CD16 exists as a polypepetide-anchored from (FCRIIIA or CD16A) on human natural killer (NK) cells and monocytes/ macrophages and as a glycosylphosphatidylinositol (GPI)-anchored form (FcRIIIB or CD16B) on neutrophils. CD16B is polymorphic and the two alleles are termed NA1 and NA2.3 CD16 plays a role in signal transduction, NK cell activation and antibody-dependent cellular cytotoxicity. This MAb has been showed to inhibit the binding of immune complex to NK cells, inhibit cytotoxicity of NK cells, and induce calcium fluxes in NK cells and neutrophils.

Type: Primary
Antigen: FCGR3A
Clonality: Monoclonal
Clone: HuNK2
Conjugation: Purified
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgG2a, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR