Anti-MITF Mouse Monoclonal Antibody (Biotin) [clone: D5 MITF/915]

Proveedor: Biotium
BNCB0950-500 BNCB0950-100
BNCB0950-500EA 560 EUR
BNCB0950-500 BNCB0950-100
Anti-MITF Mouse Monoclonal Antibody (Biotin) [clone: D5 MITF/915]
Anticuerpos
MITF (microphthalmia transcription factor) is a basic helix-loop-helix-leucine-zipper (bHLH-Zip) transcription factor that regulates the development and survival of melanocytes and retinal pigment epithelium, and also is involved in transcription of pigmentation enzyme genes such as tyrosinase TRP1 and TRP2. MITF has been shown to be phosphorylated by MAP kinase in response to c-kit activation, resulting in upregulation of MITF transcriptional activity. Mutations of the MITF gene are associated with the autosomal dominant hereditary deafness and pigmentation condition, Waardenburg Syndrome type 2A. Multiple isoforms of MITF exist, including MITF-A, MITF-B, MITF-C, MITF-H, and MITF-M, which differ in the amino-terminal domain and in their expression patterns. The MITF-M isoform is restricted to the melanocyte cell lineage. Anti-MITF, D5, recognizes a nuclear protein, which is expressed in the majority of primary and metastatic epithelioid malignant melanomas as well as in normal melanocytes, benign nevi and dysplastic nevi.

Type: Primary
Antigen: MITF
Clonality: Monoclonal
Clone: D5 MITF/915
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR