Anti-P62 Rabbit Polyclonal Antibody
Proveedor: Novus Biologicals
NBP1-42821 NBP1-42821SS
NOVUNBP1-42821EA 554 EUR
NOVUNBP1-42821 NOVUNBP1-42821SS
Anti-P62 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit Polyclonal p62/SQSTM1 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Mouse, Rat.
Type: Primary
Antigen: P62
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human