Anti-P62 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP1-42822 NBP1-42822SS
NOVUNBP1-42822EA 548 EUR
NOVUNBP1-42822 NOVUNBP1-42822SS
Anti-P62 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit Polyclonal p62/SQSTM1 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: P62
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR