Anti-CD31 Rat Monoclonal Antibody [clone: MEC 7.46]

Proveedor: Novus Biologicals
NB100-1642 NB100-1642SS
NOVUNB100-1642EA 654 EUR
NOVUNB100-1642 NOVUNB100-1642SS
Anti-CD31 Rat Monoclonal Antibody [clone: MEC 7.46]
Anticuerpos
Rat Monoclonal CD31/PECAM1 Antibody (MEC 7.46). Tested Applications: Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Mouse.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR