Anti-IGF1 Mouse Monoclonal Antibody (CF488A) [clone: IGF1/1020]

Proveedor: Biotium
BNC881020-500 BNC881020-100
BTIUBNC881020-500EA 560 EUR
BTIUBNC881020-500 BTIUBNC881020-100
Anti-IGF1 Mouse Monoclonal Antibody (CF488A) [clone: IGF1/1020]
Anticuerpos
This antibody is specific to Insulin-like Growth Factor (IGF-1) and shows minimal cross-reaction with IGF-11, Proinsulin, MSF, and Insulin. IGF-1 is a polypeptide growth factor with two isoforms that are produced by alternative splicing. Isoform 1 is also known as IGF-IB while isoform 2 is known as IGF-IA. IGF-1 stimulates the proliferation of a wide range of cell types including muscle, bone and cartilage tissue. It functions as an autocrine regulator of growth. Activation of IGF system has emerged as a key factor for tumor progression and resistance to apoptosis in many cancers like those of breast, thyroid and colon.

Type: Primary
Antigen: IGF1
Clonality: Monoclonal
Clone: IGF1/1020
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR