Anti-CD66 molecule Mouse Monoclonal Antibody (CF488A) [clone: C66/1030]

Proveedor: Biotium
BNC881030-500 BNC881030-100
BNC881030-500EA 560 EUR
BNC881030-500 BNC881030-100
Anti-CD66 molecule Mouse Monoclonal Antibody (CF488A) [clone: C66/1030]
Anticuerpos
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci ofMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno CD66 molecule
Environmentally Preferable
Nombre del antígeno CD66
Sinónimos de antígenos CEACAM5|CD66|Biliary Glycoprotein (BGP-1)|Carcinoembryonic Antigen-related Cell Adhesion Molecule 5
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 488A
Clon Clone C66/1030
Reactividad Human
Huésped Mouse
ID del gen 709196
Isotipo IgG1 kappa
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR