Anti-CD84 Mouse Monoclonal Antibody (CF640R) [clone: 153-4D9]

Proveedor: Biotium
BNC401049-500 BNC401049-100
BNC401049-500EA 560 EUR
BNC401049-500 BNC401049-100
Anti-CD84 Mouse Monoclonal Antibody (CF640R) [clone: 153-4D9]
Anticuerpos
Recognizes a protein of 74 kDa, identified as CD84. It is expressed on mature B cells and on B-cell lines, including pre-B-cell lines, but not on plasma cell lines. Immunohistochemical studies demonstrated that CD84 strongly expressed on tissues macrophages. CD84 is also highly expressed on platelets and, at low levels on peripheral blood T cells. It is a highly N-glycosylated protein and belongs to immunoglobulin superfamily. It may play a role in leukocyte activation.

Type: Primary
Antigen: CD84
Clonality: Monoclonal
Clone: 153-4D9
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno CD84
Environmentally Preferable
Sinónimos de antígenos LY9B|Signaling lymphocytic activation molecule 5 (SLAMF5)|hCD84|Cell surface antigen MAX.3|SLAM family member 5|Hly9-beta|Leucocyte differentiation antigen CD84
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 640R
Clon Clone 153-4D9
Reactividad Human
Huésped Mouse
ID del gen 398093
Isotipo IgG1 kappa
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR