Anti-CDH16 Mouse Monoclonal Antibody (CF488A) [clone: CDH16/1071]

Proveedor: Biotium
BNC881071-500 BNC881071-100
BNC881071-500EA 560 EUR
BNC881071-500 BNC881071-100
Anti-CDH16 Mouse Monoclonal Antibody (CF488A) [clone: CDH16/1071]
Anticuerpos
This MAb recognizes a protein of 130 kDa, identified as Ksp-cadherin. Cadherins form a superfamily of related glycoproteins that mediate calcium-dependent cell adhesion and transmit signals from the extracellular matrix to the cytoplasm. Cadherins have been implicated in embryogenesis, tissue morphogenesis, tissue structure maintenance, cell polarization, neoplastic invasiveness and metastasis, and membrane transport. It is suggested that Ksp-cadherin is a marker for terminal differentiation of the basolateral membranes of renal tubular epithelial cells. Within the kidney, Ksp-Cadherin is found exclusively in the basolateral membrane of renal tubular epithelial cells and collecting duct cells, and not in glomeruli, renal interstitial cells, or blood vessels.Ksp-Cadherin has been suggested to distinguish Chromophobe Renal-Cell Carcinoma from Oncocytoma.

Type: Primary
Antigen: CDH16
Clonality: Monoclonal
Clone: CDH16/1071
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Dog, Human, Mouse, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR