Anti-HLA-DRA Mouse Monoclonal Antibody (CF405S) [clone: 169-1B5.2]

Proveedor: Biotium
BNC041096-500 BNC881096-100
BNC041096-500EA 560 EUR
BNC041096-500 BNC881096-100
Anti-HLA-DRA Mouse Monoclonal Antibody (CF405S) [clone: 169-1B5.2]
Anticuerpos
This antibody detects a monomorphic general framework determinant of HLA-DR Class II antigen. It does not cross react with HLA-DP and HLA-DQ. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36 kDa alpha (heavy) chain and a 28 kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4 T cells. It is an excellent histiocytic marker in paraffin sections producing intenseMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno MHC2 MHC2
Environmentally Preferable
Nombre del antígeno MHC II MHC II
Sinónimos de antígenos HLA-DRA HLA-DRA
Tipo de anticuerpo Primario Primario
Clonalidad Monoclonal Monoclonal
Conjugado CF® 405S CF® 488A
Clon Clone 169-1B5.2 Clone 169-1B5.2
Reactividad Human,Bovine,Cat Human,Bovine,Cat
Huésped Mouse Mouse
ID del gen 520048 520048
Isotipo IgG2b kappa IgG2b kappa
FluorescenciaInmunológica Yes Yes
Concentración 0,1 mg/ml 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR