Anti-TGFA Mouse Monoclonal Antibody (Purified) [clone: TGFA/1119]

Proveedor: Biotium
BNUB1119-500 BNUB1119-100
BTIUBNUB1119-500EA 511 EUR
BTIUBNUB1119-500 BTIUBNUB1119-100
Anti-TGFA Mouse Monoclonal Antibody (Purified) [clone: TGFA/1119]
Anticuerpos
This antibody reacts with the C-terminus of TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. TGF alpha (aa50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Type: Primary
Antigen: TGFA
Clonality: Monoclonal
Clone: TGFA/1119
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR