Anti-TGFA Mouse Monoclonal Antibody (Biotin) [clone: TGFA/1119]

Proveedor: Biotium
BNCB1119-500 BNCB1119-100
BNCB1119-500EA 560 EUR
BNCB1119-500 BNCB1119-100
Anti-TGFA Mouse Monoclonal Antibody (Biotin) [clone: TGFA/1119]
Anticuerpos
This antibody reacts with the C-terminus of TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. TGF alpha (aa50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Type: Primary
Antigen: TGFA
Clonality: Monoclonal
Clone: TGFA/1119
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno TGFA
Nombre del antígeno TGFalpha
Environmentally Preferable
Sinónimos de antígenos TGFA|Waved 1|ETGF|TFGA|TGF Type 1|Wa1|EGF-like TGF
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Biotin
Clon Clone TGFA/1119
Reactividad Human
Huésped Mouse
ID del gen 170009
Isotipo IgG1 kappa
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR