Anti-TOX3 Mouse Monoclonal Antibody (CF594) [clone: TOX3/1123]

Proveedor: Biotium
BNC941123-500 BNC941123-100
BNC941123-500EA 560 EUR
BNC941123-500 BNC941123-100
Anti-TOX3 Mouse Monoclonal Antibody (CF594) [clone: TOX3/1123]
Anticuerpos
This antibody recognizes a 63 kDa protein, which is identified as TOX3. It contains a high mobility group (HMG)-box, which regulates Ca2 -dependent transcription in neurons through interaction with the cAMP-response-element-binding protein (CREB). TOX3 appears to be associated with breast cancer susceptibility and is expressed downstream of a cytoprotective cascade together with CITED1, a transcriptional regulator that does not bind directly to DNA. TOX3 is predominantly expressed in the brain and forms homodimers. TOX3 overexpression protects neuronal cells from cell death caused byMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno TOX3 TOX3
Environmentally Preferable
Nombre del antígeno TOX high mobility group box family member 3 TOX high mobility group box family member 3
Sinónimos de antígenos CAG trinucleotide repeat-containing gene F9 protein|CAGF9|Trinucleotide repeat containing 9 (TNRC9)|TOX3|TOX high mobility group box family member 3 CAG trinucleotide repeat-containing gene F9 protein|CAGF9|Trinucleotide repeat containing 9 (TNRC9)|TOX3|TOX high mobility group box family member 3
Tipo de anticuerpo Primario Primario
Clonalidad Monoclonal Monoclonal
Conjugado CF® 594 CF® 594
Clon Clone TOX3/1123 Clone TOX3/1123
Reactividad Human Human
Huésped Mouse Mouse
ID del gen 460789 460789
Isotipo IgG2b kappa IgG2b kappa
InmunoQuímica Yes Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR