Anti-MUC5AC Mouse Monoclonal Antibody (CF568) [clone: MUC5AC/917 45M1]

Proveedor: Biotium
BNC681134-500 BNC681134-100
BNC681134-500EA 560 EUR
BNC681134-500 BNC681134-100
Anti-MUC5AC Mouse Monoclonal Antibody (CF568) [clone: MUC5AC/917 45M1]
Anticuerpos
Mucin 5AC glycoprotein (MUC5AC) is a 641 kDa glycoprotein belonging to the superfamily of mucins. Mucins are high molecular weight glycoproteins produced by epithelial cells and can be divided into two families; secretory mucins and membrane bound mucins. MUC5AC is a mucus-forming secreted mucin that is found in normal gastric and tracheo-bronchial mucosa, but absent from normal colon. MUC5AC expression is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno MUC5AC
Nombre del antígeno Mucin 5AC
Environmentally Preferable
Sinónimos de antígenos Mucin 5AC oligomeric mucus/gel forming|Mucin 5 Subtypes A And C|Apomucin Major Airway Glycoprotein|Tracheobronchial Mucin (TBM)|Mucin 5 subtype AC tracheobronchial
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado CF® 568
Clon Clone MUC5AC/917 45M1
Reactividad Human
Huésped Mouse
ID del gen 534332
Isotipo IgGs
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR