Anti-CD66 molecule Mouse Monoclonal Antibody (CF488A) [clone: C66/195 C66/261 C66/1009 C66/1030]

Proveedor: Biotium
BNC881139-500 BNC881139-100
BNC881139-500EA 560 EUR
BNC881139-500 BNC881139-100
Anti-CD66 molecule Mouse Monoclonal Antibody (CF488A) [clone: C66/195 C66/261 C66/1009 C66/1030]
Anticuerpos
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci of gastric carcinoma and in distinguishing pulmonary adenocarcinomas (60-70% are CEA ) from pleural mesotheliomas (rarely or weakly CEA ). Anti-CEA positivity is seen in adenocarcinomas from the lung, colon, stomach, esophagus, pancreas, gallbadder, urachus, salivary gland, ovary, and endocervix.

Type: Primary
Antigen: CD66 molecule
Clonality: Monoclonal
Clone: C66/195 C66/261 C66/1009 C66/1030
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR